Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143667.1 | NP_001137139.1 | 3 | 602 | ||
NM_021211.3 | NP_067034.2 | 3 | 634 | |||
GenBank mRNA | AF205600.1 | - | 3 | 603 | ||
AK022942.1 | - | 1 | 779 | |||
AK025829.1 | - | 1 | 114 | |||
AK123233.1 | - | 2 | 1954 | |||
AK315033.1 | - | 3 | 566 | |||
AL137577.1 | - | 3 | 1321 | |||
BC017803.1 | - | 3 | 574 | |||
BC047754.1 | - | 3 | 602 | 6471771 |