Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021218.1 | NP_067041.1 | 5 | 630 | ||
GenBank mRNA | AF161409.1 | - | 5 | 597 | ||
AK000637.1 | - | 5 | 633 | |||
AK310007.1 | - | 4 | 539 | |||
BC014881.1 | - | 5 | 630 | 3907074 | ||
BC065210.1 | - | 5 | 628 | 3456693 |
Gene Symbol: HSPA4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011531745.2 | XP_011530047.1 | 12 | 2360 |