Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRUNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303242.1 | NP_001290171.1 | 3 | 391 | ||
NM_001303243.1 | NP_001290172.1 | 3 | 391 | |||
NM_021222.2 | NP_067045.1 | 3 | 391 | |||
GenBank mRNA | AF051907.1 | - | 3 | 305 | ||
AK315120.1 | - | 3 | 322 | |||
BC025304.1 | - | 3 | 285 | 5458046 | ||
BM554427.1 | - | 3 | 305 | |||
DA719664.1 | - | 3 | 391 | |||
U67085.1 | - | 2 | 97 |