Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001987.4 | NP_001978.1 | 3 | 521 | ||
XM_011520607.2 | XP_011518909.1 | 3 | 771 | |||
XM_011520608.2 | XP_011518910.1 | 3 | 1110 | |||
XM_011520609.2 | XP_011518911.1 | 3 | 1385 | |||
XM_011520611.2 | XP_011518913.1 | 2 | 318 | |||
XM_017018990.1 | XP_016874479.1 | 3 | 774 | |||
XM_017018991.1 | XP_016874480.1 | 7 | 1827 | |||
GenBank mRNA | AK289441.1 | - | 3 | 389 | ||
BC043399.1 | - | 3 | 521 | 6014394 | ||
FR719953.1 | - | 3 | 241 | |||
KU178049.1 | - | 3 | 247 | |||
KU178050.1 | - | 3 | 247 | |||
U11732.1 | - | 3 | 271 | |||
Z35761.1 | - | 3 | 247 |