Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021223.2 | NP_067046.1 | 4, 5 | 392 | ||
XM_011515463.2 | XP_011513765.1 | 5, 6 | 536 | |||
XM_011515464.2 | XP_011513766.1 | 5, 6 | 466 | |||
XM_017012479.1 | XP_016867968.1 | 5, 6 | 473 | |||
GenBank mRNA | AK311869.1 | - | 5, 6 | 407 | ||
BC027915.1 | - | 4, 5 | 388 | 5223298 | ||
M94547.1 | - | 4, 5 | 392 |