Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRODH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021232.1 | NP_067055.1 | 1 | 147 | ||
GenBank mRNA | U80018.1 | - | 1 | 147 |