Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021095.2 | NP_066918.2 | 9 | 1463 | ||
NR_028323.1 | 9 | 1463 | ||||
XM_006712128.1 | XP_006712191.1 | 9 | 1258 | |||
XM_006712129.1 | XP_006712192.1 | 9 | 1306 | |||
XM_006712130.1 | XP_006712193.1 | 9 | 1297 | |||
XM_011533146.2 | XP_011531448.1 | 7 | 1023 | |||
XM_017005216.1 | XP_016860705.1 | 6 | 1033 | |||
GenBank mRNA | AF069307.1 | - | 9 | 1359 | ||
AF081571.1 | - | 8 | 1175 | |||
AK027855.1 | - | 9 | 1304 | |||
AK293162.1 | - | 3 | 347 | |||
AK314545.1 | - | 10 | 1290 | |||
BC012806.2 | - | 7 | 1025 | 3507776 | ||
BC015631.2 | - | 8 | 1176 | 3628126 |