Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021101.4 | NP_066924.1 | 2 | 584 | ||
GenBank mRNA | AF101051.1 | - | 2 | 564 | ||
AF114837.1 | - | 2 | 573 | |||
AF115546.1 | - | 2 | 354 | |||
AF134160.1 | - | 2 | 591 | |||
AK296934.1 | - | 3 | 465 | |||
AK312866.1 | - | 2 | 580 | |||
AY358652.1 | - | 2 | 562 | |||
BC012471.1 | - | 2 | 542 | 4500534 | ||
CR457138.1 | - | 2 | 344 | |||
EF564137.1 | - | 2 | 344 |