Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021129.3 | NP_066952.1 | 4 | 304 | ||
GenBank mRNA | AB026723.1 | - | 4 | 227 | ||
AF092439.1 | - | 4 | 274 | |||
AF108211.1 | - | 4 | 185 | |||
AF119665.1 | - | 4 | 274 | |||
AF154065.1 | - | 4 | 242 | |||
AF217186.1 | - | 4 | 197 | |||
AK058193.1 | - | 4 | 297 | |||
AY295767.1 | - | 1 | 393 | |||
BC001022.2 | - | 4 | 279 | 3345496 | ||
BC061581.1 | - | 4 | 287 | 6526628 | ||
BC105034.1 | - | 4 | 270 | 8144037 | ||
BC105036.1 | - | 4 | 270 | 8144039 | ||
BC107882.1 | - | 4 | 238 | 6495107 | ||
BU520836.1 | - | 4 | 285 | |||
EU794626.1 | - | 4 | 304 |