Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143935.1 | NP_001137407.1 | 6 | 688 | ||
NM_021151.3 | NP_066974.2 | 5 | 604 | |||
XM_011516337.2 | XP_011514639.1 | 5 | 604 | |||
XM_017012370.1 | XP_016867859.1 | 5 | 542 | |||
GenBank mRNA | AF073770.1 | - | 5 | 495 | ||
AF168793.1 | - | 5 | 495 | |||
AK127901.1 | - | 6 | 644 | |||
AK313487.1 | - | 5 | 548 | |||
BC039004.1 | - | 5 | 544 | 5242962 |