Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 2 | 150 | ||
NM_001144894.1 | NP_001138366.1 | 2 | 150 | |||
NM_001144895.1 | NP_001138367.1 | 4 | 282 | |||
NM_001144896.1 | NP_001138368.1 | 3 | 210 | |||
NM_001144897.1 | NP_001138369.1 | 4 | 282 | |||
NM_001144899.1 | NP_001138371.1 | 4 | 282 | |||
NM_021155.3 | NP_066978.1 | 4 | 282 | |||
NR_026692.1 | 3 | 382 | ||||
XM_005272472.4 | XP_005272529.1 | 4 | 313 | |||
GenBank mRNA | AF290886.1 | - | 4 | 224 | ||
AK293089.1 | - | 4 | 238 | |||
AK313585.1 | - | 4 | 283 | |||
AY042221.1 | - | 4 | 215 | |||
AY042222.1 | - | 4 | 215 | |||
AY042223.1 | - | 4 | 215 | |||
AY042225.1 | - | 3 | 143 | |||
AY042226.1 | - | 2 | 83 | |||
AY042227.1 | - | 2 | 83 | |||
AY042229.1 | - | 3 | 315 | |||
AY042230.1 | - | 2 | 243 | |||
AY042231.1 | - | 2 | 243 | |||
AY042232.1 | - | 2 | 243 | |||
AY042233.1 | - | 2 | 243 | |||
BC110615.2 | - | 2 | 250 | 40032459 | ||
M98457.1 | - | 4 | 256 |
Gene Symbol: CLEC4M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC110614.2 | - | 2 | 224 | 40032453 |