Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021167.4 | NP_066990.3 | 2 | 630 | ||
NR_052016.1 | 2 | 630 | ||||
GenBank mRNA | AK026142.1 | - | 2 | 599 | ||
AK315788.1 | - | 2 | 596 | |||
BC019350.1 | - | 2 | 601 | 3626609 | ||
BC031091.1 | - | 2 | 504 | 5275963 |