Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM60A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135811.1 | NP_001129283.1 | 4 | 539 | ||
NM_001135812.1 | NP_001129284.1 | 3 | 435 | |||
NM_021238.2 | NP_067061.1 | 4 | 535 | |||
XM_011520803.2 | XP_011519105.1 | 3 | 537 | |||
XM_017019758.1 | XP_016875247.1 | 4 | 603 | |||
XM_017019759.1 | XP_016875248.1 | 5 | 649 | |||
XM_017019760.1 | XP_016875249.1 | 5 | 654 | |||
XM_017019761.1 | XP_016875250.1 | 4 | 584 | |||
XM_017019762.1 | XP_016875251.1 | 4 | 501 | |||
GenBank mRNA | AF087885.1 | - | 4 | 488 | ||
AF212220.1 | - | 3 | 394 | |||
AK026932.1 | - | 3 | 346 | |||
AK294457.1 | - | 3 | 448 | |||
AL157432.1 | - | 4 | 487 | |||
AY598323.1 | - | 2 | 213 | |||
BC000024.2 | - | 4 | 535 | 3504430 | ||
BC071966.1 | - | 3 | 315 | 5924274 | ||
BX648630.1 | - | 2 | 530 |