Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005207.3 | NP_005198.1 | 1, 2 | 828 | ||
GenBank mRNA | AK292909.1 | - | 2, 3 | 800 | ||
BC026248.1 | - | 1, 2 | 820 | |||
BC043500.1 | - | 1, 2 | 769 | 6046673 | ||
CR456423.1 | - | 1, 2 | 475 | |||
X59656.1 | - | 1, 2 | 828 |