Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271028.1 | NP_001257957.1 | 10 | 2022 | ||
NM_021153.3 | NP_066976.1 | 12 | 2392 | |||
NR_073130.1 | 12 | 2364 | ||||
XM_011525931.2 | XP_011524233.1 | 12 | 2359 | |||
XM_017025711.1 | XP_016881200.1 | 12 | 2281 | |||
GenBank mRNA | AF047826.1 | - | 4 | 730 | ||
AJ007607.1 | - | 12 | 2233 | |||
AK315257.1 | - | 12 | 2366 | |||
AY358654.1 | - | 11 | 2131 |