Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021191.2 | NP_067014.2 | 2 | 947 | ||
GenBank mRNA | AK314136.1 | - | 2 | 862 | ||
BC040961.1 | - | 1 | 437 | 5490979 |