Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130033.1 | NP_001123505.1 | 7 | 1601 | ||
NM_001130034.1 | NP_001123506.1 | 8 | 1447 | |||
NM_001130035.1 | NP_001123507.1 | 7 | 1342 | |||
NM_001130036.1 | NP_001123508.1 | 6 | 1474 | |||
NM_021200.2 | NP_067023.1 | 8 | 1706 | |||
XM_011545192.2 | XP_011543494.1 | 8 | 2280 | |||
XM_011545193.2 | XP_011543495.1 | 7 | 2175 | |||
GenBank mRNA | AB209716.1 | - | 7 | 1254 | ||
AF081583.1 | - | 7 | 1225 | |||
AF093249.1 | - | 7 | 1226 | |||
AF100612.1 | - | 8 | 1331 | |||
AF101054.1 | - | 7 | 1597 | |||
AK054725.1 | - | 3 | 1359 | |||
AK296033.1 | - | 7 | 1146 | |||
AL712105.1 | - | 1 | 111 | |||
BC008075.2 | - | 7 | 1280 | 3050369 | ||
U89715.1 | - | 8 | 1702 |