Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099855.1 | NP_001093325.1 | 7, 8 | 771 | ||
NM_001171876.1 | NP_001165347.1 | 7, 8 | 771 | |||
NM_001171877.1 | NP_001165348.1 | 3, 4 | 636 | |||
NM_001171878.1 | NP_001165349.1 | 4, 5 | 753 | |||
NM_001171879.1 | NP_001165350.1 | 4, 5 | 753 | |||
NM_005369.4 | NP_005360.3 | 4, 5 | 753 | |||
XM_005262413.4 | XP_005262470.1 | 8, 9 | 919 | |||
XM_011531339.2 | XP_011529641.1 | 8, 9 | 978 | |||
XM_017029529.1 | XP_016885018.1 | 7, 8 | 740 | |||
XM_017029530.1 | XP_016885019.1 | 7, 8 | 740 | |||
XM_017029531.1 | XP_016885020.1 | 7, 8 | 673 | |||
XM_017029532.1 | XP_016885021.1 | 4, 5 | 830 | |||
GenBank mRNA | AB085901.1 | - | 4, 5 | 430 | ||
AB085902.1 | - | 4, 5 | 430 | |||
AK296488.1 | - | 3, 4 | 636 | |||
AK296529.1 | - | 7, 8 | 871 | |||
AK302957.1 | - | 7, 8 | 719 | |||
AK313900.1 | - | 4, 5 | 613 | |||
AK316416.1 | - | 7, 8 | 759 | |||
AL117234.1 | - | 6, 7 | 654 | |||
DA326503.1 | - | 4, 5 | 575 | |||
X12556.1 | - | 4, 5 | 604 |