Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256410.1 | NP_001243339.1 | 8 | 2138 | ||
NM_001256411.1 | NP_001243340.1 | 6 | 1984 | |||
NM_001256412.1 | NP_001243341.1 | 5 | 1859 | |||
NM_001256415.1 | NP_001243344.1 | 6 | 1979 | |||
NM_021252.4 | NP_067075.1 | 7 | 2051 | |||
NR_046172.1 | 6 | 1989 | ||||
GenBank mRNA | AB209698.1 | - | 3 | 6813 | ||
AK001555.1 | - | 7 | 1855 | |||
AL136734.1 | - | 7 | 1904 | |||
AY574034.1 | - | 8 | 2027 | |||
BC015014.2 | - | 7 | 1905 | 3922029 | ||
BC029350.1 | - | 7 | 1921 | 4819866 |