Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134647.1 | NP_001128119.1 | 2 | 355 | ||
NM_198595.2 | NP_940997.1 | 2 | 355 | |||
XM_006713909.3 | XP_006713972.1 | 2 | 238 | |||
XM_011513544.2 | XP_011511846.1 | 2 | 408 | |||
XM_017008535.1 | XP_016864024.1 | 1 | 84 | |||
XM_017008536.1 | XP_016864025.1 | 1 | 84 | |||
GenBank mRNA | AF188700.1 | - | 1 | 82 | ||
AK290807.1 | - | 2 | 182 | |||
AK297631.1 | - | 2 | 148 | |||
AL706541.1 | - | 1 | 300 | |||
BC032777.1 | - | 1 | 106 | 4914388 | ||
DA605903.1 | - | 2 | 288 | |||
DB450895.1 | - | 2 | 355 |