Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGIRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135053.1 | NP_001128525.1 | 10 | 1486 | ||
NM_001135054.1 | NP_001128526.1 | 10 | 1559 | |||
NM_021805.2 | NP_068577.2 | 10 | 1455 | |||
XM_005253044.2 | XP_005253101.1 | 9 | 1848 | |||
XM_005253045.1 | XP_005253102.1 | 9 | 1744 | |||
XM_005253046.1 | XP_005253103.1 | 9 | 1832 | |||
XM_005253048.4 | XP_005253105.1 | 8 | 1456 | |||
XM_017018099.1 | XP_016873588.1 | 10 | 1766 | |||
XM_017018100.1 | XP_016873589.1 | 9 | 1595 | |||
GenBank mRNA | AK025099.1 | - | 10 | 1455 | ||
AK093427.1 | - | 7 | 1683 | |||
AK172830.1 | - | 10 | 1455 | |||
AY358342.1 | - | 8 | 1621 | |||
BC003591.1 | - | 9 | 1385 | 2821373 | ||
BC025953.1 | - | 10 | 1414 | 3939162 | ||
BC106007.1 | - | 10 | 1536 | 6092503 | ||
BX369506.2 | - | 5 | 654 |