Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171132.2 | NP_001164603.1 | 5 | 545 | ||
NM_001171133.2 | NP_001164604.1 | 4 | 431 | |||
NM_001171134.2 | NP_001164605.1 | 4 | 642 | |||
NM_001282311.1 | NP_001269240.1 | 5 | 740 | |||
NM_001282312.1 | NP_001269241.1 | 5 | 486 | |||
NM_021806.3 | NP_068578.2 | 4 | 642 | |||
XM_005274714.3 | XP_005274771.1 | 6 | 590 | |||
XM_005274716.3 | XP_005274773.1 | 6 | 590 | |||
XM_005277879.4 | XP_005277936.1 | 5 | 551 | |||
XM_006724831.2 | XP_006724894.1 | 6 | 590 | |||
XM_006724832.3 | XP_006724895.1 | 5 | 551 | |||
XM_006724833.2 | XP_006724896.1 | 6 | 590 | |||
XM_006724834.2 | XP_006724897.1 | 5 | 703 | |||
XM_011531185.2 | XP_011529487.1 | 3 | 148 | |||
XM_017029699.1 | XP_016885188.1 | 4 | 863 | |||
XM_017029700.1 | XP_016885189.1 | 4 | 862 | |||
XM_017029701.1 | XP_016885190.1 | 5 | 765 | |||
XM_017029702.1 | XP_016885191.1 | 4 | 378 | |||
GenBank mRNA | AK024601.1 | - | 5 | 502 | ||
AK124890.1 | - | 1 | 2833 | |||
AK294116.1 | - | 4 | 605 | |||
AK314682.1 | - | 4 | 560 | |||
AL539139.3 | - | 5 | 737 | |||
BC002934.1 | - | 5 | 644 | |||
BC008912.2 | - | 4 | 293 | 3639043 | ||
BM457676.1 | - | 5 | 527 | |||
CX867071.1 | - | 4 | 349 | |||
X87193.1 | - | 4 | 449 |