Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170794.1 | NP_001164265.1 | 4 | 810 | ||
NM_021813.3 | NP_068585.1 | 6 | 981 | |||
XM_005248758.4 | XP_005248815.1 | 3 | 1763 | |||
XM_005248759.4 | XP_005248816.1 | 2 | 1683 | |||
XM_011536039.2 | XP_011534341.1 | 5 | 892 | |||
XM_011536040.2 | XP_011534342.1 | 4 | 841 | |||
XM_017011156.1 | XP_016866645.1 | 9 | 1953 | |||
XM_017011157.1 | XP_016866646.1 | 8 | 1759 | |||
XM_017011158.1 | XP_016866647.1 | 10 | 2032 | |||
XM_017011159.1 | XP_016866648.1 | 11 | 2121 | |||
XM_017011160.1 | XP_016866649.1 | 8 | 1875 | |||
XM_017011161.1 | XP_016866650.1 | 10 | 1583 | |||
XM_017011162.1 | XP_016866651.1 | 7 | 1782 | |||
XM_017011163.1 | XP_016866652.1 | 9 | 1954 | |||
XM_017011164.1 | XP_016866653.1 | 8 | 1860 | |||
XM_017011165.1 | XP_016866654.1 | 5 | 907 | |||
XM_017011166.1 | XP_016866655.1 | 7 | 1065 | |||
XM_017011167.1 | XP_016866656.1 | 9 | 1953 | |||
GenBank mRNA | AB208889.1 | - | 4 | 447 | ||
AF357835.1 | - | 3 | 306 | |||
AJ271878.1 | - | 6 | 815 | |||
AK309526.1 | - | 6 | 1112 | |||
HE578163.1 | - | 1 | 75 |