Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021255.2 | NP_067078.1 | 2 | 410 | ||
XM_005267890.4 | XP_005267947.1 | 2 | 455 | |||
XM_011536990.2 | XP_011535292.1 | 3 | 120 | |||
XM_011536992.2 | XP_011535294.1 | 2 | 173 | |||
XM_017021478.1 | XP_016876967.1 | 2 | 802 | |||
XM_017021479.1 | XP_016876968.1 | 3 | 9352 | |||
GenBank mRNA | AF302502.1 | - | 2 | 300 | ||
AK315727.1 | - | 2 | 372 | |||
BC009476.2 | - | 2 | 410 | 3942712 |