Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021627.2 | NP_067640.2 | 3, 4 | 470 | ||
XM_005247690.2 | XP_005247747.2 | 3, 4 | 485 | |||
GenBank mRNA | AF151697.2 | - | 3, 4 | 473 | ||
AK027599.1 | - | 3, 4 | 411 | |||
AK298628.1 | - | 3, 4 | 297 | |||
AK304402.1 | - | 3, 4 | 539 | |||
AL834380.1 | - | 3, 4 | 294 | |||
BC040609.1 | - | 3, 4 | 356 | 5163937 |