Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005687.4 | NP_005678.3 | 3 | 456 | ||
NR_130154.1 | 4 | 652 | ||||
XM_006712169.2 | XP_006712232.1 | 4 | 358 | |||
XM_011510466.2 | XP_011508768.1 | 4 | 230 | |||
XM_017003110.1 | XP_016858599.1 | 4 | 2631 | |||
GenBank mRNA | AF042346.1 | - | 3 | 174 | ||
AF161521.1 | - | 3 | 194 | |||
AK055289.1 | - | 1 | 1420 | |||
AK294158.1 | - | 4 | 212 | |||
BC006502.2 | - | 3 | 163 | |||
BC017783.1 | - | 3 | 186 | 4664949 | ||
BG721840.1 | - | 3 | 458 | |||
D84430.1 | - | 3 | 187 |