Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242828.1 | NP_001229757.1 | 3 | 540 | ||
NM_001242830.1 | NP_001229759.1 | 3 | 540 | |||
NM_001301856.1 | NP_001288785.1 | 3 | 773 | |||
NM_021814.4 | NP_068586.1 | 3 | 540 | |||
GenBank mRNA | AB209798.1 | - | 3 | 288 | ||
AF231981.1 | - | 2 | 169 | |||
AF338241.1 | - | 3 | 513 | |||
AK125098.1 | - | 3 | 617 | |||
AK299674.1 | - | 3 | 311 | |||
AL136939.1 | - | 3 | 288 | |||
BC067123.1 | - | 3 | 289 | 6160926 | ||
DC345924.1 | - | 3 | 541 |