Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282721.1 | NP_001269650.1 | 2 | 111 | ||
NM_001282723.1 | NP_001269652.1 | 3 | 267 | |||
NM_021925.3 | NP_068744.1 | 3 | 267 | |||
NR_104233.1 | 2 | 111 | ||||
XM_011533014.2 | XP_011531316.1 | 3 | 294 | |||
GenBank mRNA | AK025473.1 | - | 3 | 205 | ||
AK299244.1 | - | 3 | 208 | |||
AK300494.1 | - | 2 | 63 | |||
AK316162.1 | - | 2 | 70 | |||
BC017473.1 | - | 3 | 224 | 3611020 |