Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286134.1 | NP_001273063.1 | 8, 9 | 2693 | ||
NM_021932.5 | NP_068751.4 | 8, 9 | 2711 | |||
XM_005253052.3 | XP_005253109.1 | 8, 9 | 1879 | |||
XM_017018107.1 | XP_016873596.1 | 8, 9 | 1963 | |||
GenBank mRNA | AK022870.1 | - | 7, 8 | 1248 | ||
AK026897.1 | - | 8, 9 | 2692 | |||
AK027090.1 | - | 5, 6 | 620 | |||
AK027461.1 | - | 8, 9 | 1381 | |||
AK225420.1 | - | 8, 9 | 1379 | |||
AK314568.1 | - | 8, 9 | 1400 | |||
AL136935.1 | - | 8, 9 | 1692 | |||
AL390088.1 | - | 8, 9 | 1414 | |||
BC011821.1 | - | 6, 7 | 1157 | |||
BC111499.1 | - | 8, 9 | 1507 | 6526180 | ||
BC121807.1 | - | 8, 9 | 1587 | 40108083 | ||
BC121808.1 | - | 8, 9 | 1587 | 40108080 | ||
BX538115.1 | - | 6, 7 | 1555 |