Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEFSEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021937.4 | NP_068756.2 | 2 | 530 | ||
XM_005247696.2 | XP_005247753.1 | 2 | 531 | |||
XM_017006975.1 | XP_016862464.1 | 2 | 487 | |||
XM_017006976.1 | XP_016862465.1 | 2 | 487 | |||
XM_017006977.1 | XP_016862466.1 | 2 | 487 | |||
XM_017006978.1 | XP_016862467.1 | 3 | 310 | |||
XM_017006979.1 | XP_016862468.1 | 2 | 799 | |||
XM_017006980.1 | XP_016862469.1 | 2 | 531 | |||
GenBank mRNA | AF268872.1 | - | 2 | 269 | ||
BC007933.2 | - | 2 | 408 | 4120858 | ||
BG420135.1 | - | 2 | 517 |