Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172673.1 | NP_001166144.1 | 11 | 1218 | ||
NM_021938.3 | NP_068757.2 | 11 | 1335 | |||
NR_033342.1 | 11 | 1132 | ||||
XM_006722832.1 | XP_006722895.1 | 11 | 1362 | |||
XM_006722833.1 | XP_006722896.1 | 11 | 1359 | |||
XM_006722836.2 | XP_006722899.1 | 10 | 1003 | |||
XM_006722837.3 | XP_006722900.1 | 11 | 1274 | |||
XM_006722838.3 | XP_006722901.1 | 10 | 1146 | |||
XM_011528173.1 | XP_011526475.1 | 10 | 975 | |||
XM_011528174.2 | XP_011526476.1 | 10 | 1144 | |||
XM_017027111.1 | XP_016882600.1 | 9 | 1218 | |||
GenBank mRNA | AB209067.1 | - | 11 | 1218 | ||
AF248649.1 | - | 1 | 114 | |||
AF329266.1 | - | 11 | 1298 | |||
AK294109.1 | - | 11 | 1132 | |||
AL832550.1 | - | 9 | 881 | |||
BC028101.1 | - | 11 | 1335 | 5242909 |