Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197104.1 | NP_001184033.1 | 2, 3 | 522 | ||
NM_005933.3 | NP_005924.2 | 2, 3 | 522 | |||
XM_011542829.2 | XP_011541131.1 | 3, 4 | 613 | |||
XM_011542830.2 | XP_011541132.1 | 3, 4 | 613 | |||
XM_011542831.2 | XP_011541133.1 | 3, 4 | 613 | |||
GenBank mRNA | AF024540.1 | - | 141 | |||
AY187942.1 | - | 178 | ||||
AY187944.1 | - | 307 | ||||
D14540.1 | - | 2, 3 | 522 | |||
L04284.1 | - | 2, 3 | 499 | |||
L04731.1 | - | 2, 3 | 313 |