Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 12 | 1476 | ||
NM_199053.2 | NP_951008.1 | 12 | 1476 | |||
XM_017008537.1 | XP_016864026.1 | 12 | 1476 | |||
XM_017008538.1 | XP_016864027.1 | 12 | 1476 | |||
XM_017008539.1 | XP_016864028.1 | 10 | 1127 | |||
GenBank mRNA | AK091146.1 | - | 7 | 700 | ||
AK096345.1 | - | 12 | 1450 | |||
AK315057.1 | - | 7 | 682 | |||
AL136752.1 | - | 4 | 385 | |||
AL833451.1 | - | 11 | 1284 | |||
AL833571.1 | - | 12 | 1455 | |||
BC051724.1 | - | 12 | 1420 | 5265781 | ||
BX538044.1 | - | 12 | 1430 | |||
BX647127.1 | - | 12 | 1447 | |||
BX648499.1 | - | 12 | 1451 | |||
CR533553.1 | - | 4 | 320 |