Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184772.2 | NP_001171701.1 | 12 | 5050 | ||
NM_021946.4 | NP_068765.3 | 11 | 4828 | |||
XM_005262452.4 | XP_005262509.2 | 14 | 5466 | |||
XM_005262453.4 | XP_005262510.2 | 14 | 5234 | |||
XM_005262454.3 | XP_005262511.2 | 13 | 5178 | |||
XM_005262455.4 | XP_005262512.2 | 13 | 5121 | |||
XM_005262456.4 | XP_005262513.2 | 13 | 5076 | |||
XM_006724776.3 | XP_006724839.1 | 14 | 5251 | |||
XM_006724777.3 | XP_006724840.1 | 13 | 5146 | |||
XM_006724779.2 | XP_006724842.1 | 14 | 5337 | |||
XM_017029721.1 | XP_016885210.1 | 14 | 5497 | |||
XM_017029722.1 | XP_016885211.1 | 13 | 5551 | |||
GenBank mRNA | AK021424.1 | - | 5 | 656 | ||
AK024395.1 | - | 1 | 287 | |||
AK074089.1 | - | 11 | 4741 | |||
AK124676.1 | - | 7 | 1214 | |||
BC044652.1 | - | 6 | 836 |