Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005934.3 | NP_005925.2 | 4 | 459 | ||
XM_011528021.1 | XP_011526323.1 | 4 | 433 | |||
XM_011528022.1 | XP_011526324.1 | 4 | 316 | |||
XM_011528023.1 | XP_011526325.1 | 4 | 433 | |||
XM_017026819.1 | XP_016882308.1 | 4 | 1015 | |||
GenBank mRNA | D14539.1 | - | 4 | 459 | ||
L04285.1 | - | 3 | 295 |