Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303051.1 | NP_001289980.1 | 12 | 3089 | ||
NM_001303109.1 | NP_001290038.1 | 10 | 2806 | |||
NM_021633.3 | NP_067646.1 | 12 | 3109 | |||
XM_011509835.1 | XP_011508137.1 | 13 | 3129 | |||
XM_011509836.1 | XP_011508138.1 | 12 | 2942 | |||
XM_011509837.1 | XP_011508139.1 | 11 | 2826 | |||
XM_017001995.1 | XP_016857484.1 | 11 | 2891 | |||
GenBank mRNA | AK024412.1 | - | 6 | 2016 | ||
AK027656.1 | - | 12 | 3080 | |||
AK130662.1 | - | 2 | 1498 |