Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134647.1 | NP_001128119.1 | 18 | 7622 | ||
NM_198595.2 | NP_940997.1 | 17 | 7370 | |||
XM_006713909.3 | XP_006713972.1 | 17 | 7253 | |||
XM_011513544.2 | XP_011511846.1 | 18 | 7675 | |||
XM_017008535.1 | XP_016864024.1 | 17 | 7153 | |||
GenBank mRNA | D25248.1 | - | 2 | 4947 |