Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136219.1 | NP_001129691.1 | 7 | 2263 | ||
NM_021642.3 | NP_067674.2 | 7 | 2260 | |||
XM_011509291.1 | XP_011507593.1 | 6 | 2210 | |||
XM_017000667.1 | XP_016856156.1 | 6 | 2125 | |||
XM_017000668.1 | XP_016856157.1 | 6 | 2122 | |||
GenBank mRNA | J03619.1 | - | 7 | 2214 | ||
M28697.1 | - | 7 | 2194 | |||
M31932.1 | - | 7 | 2212 | |||
X62572.1 | - | 7 | 2197 |