Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005937.3 | NP_005928.2 | 4 | 303 | ||
XM_006721911.2 | XP_006721974.1 | 4 | 303 | |||
XM_017024649.1 | XP_016880138.1 | 4 | 303 | |||
XM_017024650.1 | XP_016880139.1 | 4 | 303 | |||
GenBank mRNA | AB209633.1 | - | 4 | 278 | ||
AK310435.1 | - | 4 | 303 | |||
AK310437.1 | - | 4 | 303 | |||
BC064612.1 | - | 4 | 391 | |||
U07932.1 | - | 4 | 290 |