Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL21R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021798.3 | NP_068570.1 | 7, 8 | 913 | ||
NM_181078.2 | NP_851564.1 | 7, 8 | 1266 | |||
NM_181079.4 | NP_851565.4 | 8, 9 | 1407 | |||
XM_011545857.2 | XP_011544159.1 | 8, 9 | 1443 | |||
XM_011545858.2 | XP_011544160.1 | 7, 8 | 1252 | |||
XM_017023257.1 | XP_016878746.1 | 8, 9 | 1396 | |||
GenBank mRNA | AF254067.1 | - | 7, 8 | 861 | ||
AF269133.1 | - | 7, 8 | 929 | |||
AK292663.1 | - | 7, 8 | 1266 | |||
AK312825.1 | - | 7, 8 | 1255 | |||
AY358826.1 | - | 7, 8 | 896 | |||
BC004348.1 | - | 7, 8 | 878 | 3634520 |