Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171132.2 | NP_001164603.1 | 9 | 948 | ||
NM_001171133.2 | NP_001164604.1 | 8 | 834 | |||
NM_001171134.2 | NP_001164605.1 | 7 | 994 | |||
NM_001282311.1 | NP_001269240.1 | 9 | 1143 | |||
NM_001282312.1 | NP_001269241.1 | 9 | 889 | |||
NM_021806.3 | NP_068578.2 | 8 | 1045 | |||
XM_005274714.3 | XP_005274771.1 | 10 | 1014 | |||
XM_005274716.3 | XP_005274773.1 | 10 | 993 | |||
XM_005277879.4 | XP_005277936.1 | 8 | 903 | |||
XM_006724831.2 | XP_006724894.1 | 10 | 1017 | |||
XM_006724832.3 | XP_006724895.1 | 9 | 978 | |||
XM_006724833.2 | XP_006724896.1 | 9 | 942 | |||
XM_006724834.2 | XP_006724897.1 | 9 | 1130 | |||
XM_011531185.2 | XP_011529487.1 | 7 | 575 | |||
XM_017029699.1 | XP_016885188.1 | 8 | 1290 | |||
XM_017029700.1 | XP_016885189.1 | 8 | 1265 | |||
XM_017029701.1 | XP_016885190.1 | 9 | 1168 | |||
XM_017029702.1 | XP_016885191.1 | 8 | 781 | |||
GenBank mRNA | AK024601.1 | - | 9 | 906 | ||
AK057945.1 | - | 9 | 763 | |||
AK124890.1 | - | 5 | 3236 | |||
AK294116.1 | - | 7 | 957 | |||
AK314682.1 | - | 8 | 963 | |||
BC002934.1 | - | 9 | 1047 | |||
BC008912.2 | - | 8 | 696 | 3639043 | ||
CX867071.1 | - | 8 | 752 | |||
X87193.1 | - | 8 | 852 |