Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170794.1 | NP_001164265.1 | 7 | 7202 | ||
NM_021813.3 | NP_068585.1 | 9 | 7373 | |||
XM_005248758.4 | XP_005248815.1 | 6 | 8155 | |||
XM_005248759.4 | XP_005248816.1 | 5 | 8075 | |||
XM_017011156.1 | XP_016866645.1 | 12 | 8345 | |||
XM_017011157.1 | XP_016866646.1 | 11 | 8151 | |||
XM_017011158.1 | XP_016866647.1 | 13 | 8424 | |||
XM_017011159.1 | XP_016866648.1 | 14 | 8513 | |||
XM_017011160.1 | XP_016866649.1 | 11 | 8267 | |||
XM_017011161.1 | XP_016866650.1 | 13 | 7975 | |||
XM_017011162.1 | XP_016866651.1 | 10 | 8174 | |||
XM_017011163.1 | XP_016866652.1 | 12 | 8346 | |||
XM_017011164.1 | XP_016866653.1 | 11 | 8252 | |||
XM_017011166.1 | XP_016866655.1 | 10 | 7457 | |||
GenBank mRNA | AJ271878.1 | - | 9 | 7214 | ||
AL512739.1 | - | 1 | 1420 |