Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000509.5 | NP_000500.2 | 7 | 925 | ||
NM_021870.2 | NP_068656.2 | 7 | 867 | |||
GenBank mRNA | AK289422.1 | - | 7 | 770 | ||
AK290824.1 | - | 7 | 816 | |||
BC007044.1 | - | 7 | 772 | 3934943 | ||
BC021674.1 | - | 7 | 774 | 3935073 | ||
BT007081.1 | - | 7 | 726 | |||
BX538098.1 | - | 6 | 614 | |||
X51473.1 | - | 5 | 504 |