Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136007.1 | NP_001129479.1 | 11 | 869 | ||
NM_001136011.1 | NP_001129483.1 | 10 | 747 | |||
NM_001136012.1 | NP_001129484.1 | 8 | 741 | |||
NM_005971.3 | NP_005962.1 | 9 | 663 | |||
NM_021910.2 | NP_068710.1 | 9 | 825 | |||
XM_005258996.4 | XP_005259053.1 | 7 | 646 | |||
XM_011527032.2 | XP_011525334.1 | 10 | 954 | |||
XM_017026877.1 | XP_016882366.1 | 9 | 613 | |||
XM_017026878.1 | XP_016882367.1 | 8 | 533 | |||
GenBank mRNA | BC005238.1 | - | 8 | 477 | 3997821 | |
BC090044.1 | - | 9 | 612 | 30395606 | ||
BE793135.1 | - | 7 | 438 | |||
BI715233.1 | - | 8 | 453 | |||
BM549935.1 | - | 6 | 553 | |||
BQ676323.1 | - | 10 | 698 | |||
BU185141.1 | - | 9 | 550 | |||
CB128589.1 | - | 7 | 554 | |||
U28249.1 | - | 9 | 752 | |||
X93036.1 | - | 8 | 480 |