Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282719.1 | NP_001269648.1 | 5, 6 | 749 | ||
NM_001282720.1 | NP_001269649.1 | 5, 6 | 731 | |||
NM_001282721.1 | NP_001269650.1 | 5, 6 | 719 | |||
NM_001282722.1 | NP_001269651.1 | 4, 5 | 575 | |||
NM_001282723.1 | NP_001269652.1 | 5, 6 | 640 | |||
NM_001282724.1 | NP_001269653.1 | 4, 5 | 514 | |||
NM_021925.3 | NP_068744.1 | 6, 7 | 875 | |||
NR_104233.1 | 4, 5 | 484 | ||||
XM_011533014.2 | XP_011531316.1 | 6, 7 | 902 | |||
GenBank mRNA | AK021672.1 | - | 2, 3 | 97 | ||
AK025473.1 | - | 6, 7 | 813 | |||
AK299244.1 | - | 5, 6 | 581 | |||
AK299555.1 | - | 5, 6 | 720 | |||
AK300494.1 | - | 4, 5 | 436 | |||
AK301493.1 | - | 4, 5 | 485 | |||
AK316162.1 | - | 5, 6 | 678 | |||
AK316204.1 | - | 4, 5 | 546 | |||
AK316310.1 | - | 5, 6 | 702 | |||
BC017473.1 | - | 6, 7 | 832 | 3611020 |