Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021930.4 | NP_068749.3 | 3, 4 | 512 | ||
XM_017012497.1 | XP_016867986.1 | 3, 4 | 387 | |||
XM_017012498.1 | XP_016867987.1 | 3, 4 | 387 | |||
GenBank mRNA | AF317622.1 | - | 3, 4 | 391 | ||
AK055508.1 | - | 3, 4 | 427 | |||
BC068483.1 | - | 3, 4 | 513 | 5270834 |