Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044305.2 | NP_001037770.1 | 11 | 1983 | ||
NM_001281439.1 | NP_001268368.1 | 11 | 1990 | |||
NM_001281440.1 | NP_001268369.1 | 11 | 1799 | |||
NM_021940.4 | NP_068759.2 | 10 | 1902 | |||
XM_011536046.2 | XP_011534348.1 | 12 | 1885 | |||
XM_011536048.2 | XP_011534350.1 | 12 | 1872 | |||
XM_017011170.1 | XP_016866659.1 | 12 | 2068 | |||
GenBank mRNA | AF442495.1 | - | 10 | 1800 | ||
AK023221.1 | - | 11 | 1808 | |||
AK074340.1 | - | 3 | 817 | |||
AK124239.1 | - | 12 | 1916 | |||
AL833906.1 | - | 4 | 991 | |||
AY055003.1 | - | 11 | 1798 | |||
AY055004.1 | - | 10 | 1717 | |||
BC028074.1 | - | 10 | 1774 | 5215869 | ||
BC036123.1 | - | 11 | 1836 | 5313112 |