Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 14 | 1593 | ||
NM_199053.2 | NP_951008.1 | 14 | 1593 | |||
XM_017008537.1 | XP_016864026.1 | 14 | 1593 | |||
XM_017008538.1 | XP_016864027.1 | 14 | 1593 | |||
XM_017008539.1 | XP_016864028.1 | 12 | 1244 | |||
GenBank mRNA | AK096345.1 | - | 14 | 1567 | ||
AK315057.1 | - | 9 | 799 | |||
AL136752.1 | - | 6 | 502 | |||
AL833451.1 | - | 13 | 1401 | |||
AL833571.1 | - | 14 | 1572 | |||
BX538044.1 | - | 14 | 1547 | |||
BX647127.1 | - | 14 | 1564 | |||
BX648499.1 | - | 14 | 1568 | |||
CR533553.1 | - | 6 | 437 |