Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021943.2 | NP_068762.1 | 6 | 1029 | ||
XM_005249269.1 | XP_005249326.1 | 5 | 998 | |||
XM_011514790.1 | XP_011513092.1 | 5 | 896 | |||
XM_017011171.1 | XP_016866660.1 | 5 | 1002 | |||
GenBank mRNA | AK023284.1 | - | 6 | 703 | ||
AK310757.1 | - | 2 | 293 | |||
BC031481.2 | - | 6 | 645 | 4448968 |