Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286561.1 | NP_001273490.1 | 7 | 702 | ||
NM_006185.3 | NP_006176.2 | 6 | 604 | |||
NR_104476.1 | 6 | 523 | ||||
XM_006718564.1 | XP_006718627.1 | 5 | 419 | |||
XM_011545054.1 | XP_011543356.1 | 5 | 419 | |||
XM_011545055.1 | XP_011543357.1 | 6 | 597 | |||
XM_011545056.2 | XP_011543358.1 | 7 | 591 | |||
XM_011545057.2 | XP_011543359.1 | 7 | 581 | |||
XM_011545058.1 | XP_011543360.1 | 5 | 618 | |||
XM_011545059.2 | XP_011543361.1 | 7 | 583 | |||
XM_011545060.2 | XP_011543362.1 | 7 | 577 | |||
XM_011545061.2 | XP_011543363.1 | 5 | 437 | |||
XM_011545062.2 | XP_011543364.1 | 5 | 450 | |||
XM_011545063.2 | XP_011543365.1 | 5 | 417 | |||
XM_011545064.1 | XP_011543366.1 | 5 | 404 | |||
XM_011545065.1 | XP_011543367.1 | 5 | 419 | |||
XM_011545066.1 | XP_011543368.1 | 5 | 419 | |||
XM_017017830.1 | XP_016873319.1 | 6 | 459 | |||
XM_017017831.1 | XP_016873320.1 | 5 | 455 | |||
GenBank mRNA | AB210007.1 | - | 6 | 405 | ||
AB621824.1 | - | 7 | 586 | |||
AK290509.1 | - | 5 | 419 | |||
AK307577.1 | - | 6 | 422 | |||
AK309041.1 | - | 5 | 426 | |||
BC004165.2 | - | 6 | 438 | |||
BC027493.1 | - | 6 | 405 | |||
BC043499.1 | - | 6 | 405 | |||
BC103765.1 | - | 6 | 523 | |||
Z11583.1 | - | 6 | 422 | |||
Z11584.1 | - | 7 | 524 |